Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL11RA
Entrez Gene ID
693251
Gene Name
interleukin 11 receptor subunit alpha
Gene Aliases
-
Chromosome Location
Chr.Z: 8328383 - 8349089 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044674.1 NP_001038139.1 7 - 8 769 72
XM_015277359.1 XP_015132845.1 7 - 8 928 72
XM_015277360.2 XP_015132846.1 7 - 8 860 72
XM_015277361.2 XP_015132847.1 7 - 8 764 72
XM_015277362.2 XP_015132848.1 7 - 8 703 72
XM_015277363.2 XP_015132849.1 7 - 8 753 72
GenBank mRNA AB161354.1 - 7 - 8 769 72

View Full Product Details