Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH19
Entrez Gene ID
422263
Gene Name
protocadherin 19
Gene Aliases
protocadherin-19
Chromosome Location
Chr.4: 5208129 - 5260666 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098607.1 NP_001092077.1 2 - 3 2475 76
XM_015278277.1 XP_015133763.1 2 - 3 2785 76
XM_015278278.2 XP_015133764.1 2 - 3 2785 76
GenBank mRNA EF175383.1 - 2 - 3 2475 76
HM056232.1 - 2 - 3 458 76
HM056233.1 - 2 - 3 458 76
HM056234.1 - 2 - 3 458 76

View Full Product Details