Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFRSF10B
Entrez Gene ID
373904
Gene Name
tumor necrosis factor receptor superfamily, member 10b
Gene Aliases
TVB
Chromosome Location
Chr.22: 1415452 - 1421965 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204115.2 NP_989446.2 9 - 10 916 71
XM_015297294.2 XP_015152780.1 9 - 10 1031 71
XM_015297295.2 XP_015152781.1 8 - 9 902 71
GenBank mRNA AF161712.1 - 9 - 10 884 71
AF161713.1 - 9 - 10 916 71
AF507016.1 - 9 - 10 827 71
KU522456.1 - 9 - 10 815 71

View Full Product Details