Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP4
Entrez Gene ID
374165
Gene Name
fatty acid binding protein 4
Gene Aliases
A-FABP, AFABP, FABP
Chromosome Location
Chr.2: 121350127 - 121353349 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204290.1 NP_989621.1 1 - 2 110 84
GenBank mRNA AF432506.1 - 1 - 2 110 84

View Full Product Details