Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3GL1
Entrez Gene ID
395202
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
Gene Aliases
SH3P8
Chromosome Location
Chr.28: 2578652 - 2605488 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204529.1 NP_989860.1 2 - 3 114 68
XM_015299915.2 XP_015155401.1 2 - 3 438 68
XM_015299916.2 XP_015155402.1 2 - 3 177 68
XM_015299917.1 XP_015155403.1 2 - 3 150 68
GenBank mRNA AJ439351.1 - 2 - 3 114 68
BX929897.1 - 2 - 3 128 68

View Full Product Details