Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPM1
Entrez Gene ID
396203
Gene Name
nucleophosmin
Gene Aliases
NO38
Chromosome Location
Chr.13: 4020680 - 4031793 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205267.1 NP_990598.1 3 - 4 320 115
GenBank mRNA BX932398.1 - 3 - 4 310 115
BX935061.2 - 3 - 4 303 115
CR732944.1 - 3 - 4 301 115
X17200.1 - - 320 115

View Full Product Details