Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA9
Entrez Gene ID
415540
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C10H15ORF44, C15orf44, INTS14
Chromosome Location
Chr.10: 18494771 - 18505021 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005825.1 NP_001005825.1 2 - 3 292 107
XM_015292093.2 XP_015147579.1 2 - 3 369 107
GenBank mRNA AJ720263.1 - 2 - 3 292 107

View Full Product Details