Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA3
Entrez Gene ID
423542
Gene Name
proteasome subunit alpha 3
Gene Aliases
-
Chromosome Location
Chr.5: 55465942 - 55474509 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006491.1 NP_001006491.1 8 - 9 652 98
GenBank mRNA AJ719752.1 - 8 - 9 652 98

View Full Product Details