Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUTC
Entrez Gene ID
423829
Gene Name
cutC copper transporter
Gene Aliases
-
Chromosome Location
Chr.6: 23054104 - 23057842 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006503.1 NP_001006503.1 3 - 4 190 89
XM_025151603.1 XP_025007371.1 3 - 4 203 89
GenBank mRNA AJ720183.1 - 3 - 4 190 89

View Full Product Details