Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RP2
Entrez Gene ID
418675
Gene Name
retinitis pigmentosa 2 (X-linked recessive)
Gene Aliases
-
Chromosome Location
Chr.1: 131504405 - 131522593 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008680.1 NP_001008680.1 4 - 5 1141 80
GenBank mRNA AJ721100.1 - 4 - 5 1141 80

View Full Product Details