Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMB2
Entrez Gene ID
417051
Gene Name
major histocompatibility complex, class II, DM beta 2
Gene Aliases
B-DMB2, B-MA2, BMA2, HLA-DMB
Chromosome Location
Chr.16: 2589313 - 2592336 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135166.1 NP_001128638.1 1 - 2 42 57

View Full Product Details