Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTP1
Entrez Gene ID
548044
Gene Name
tyrosine phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.10: 44432853 - 44437744 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250991.1 NP_001237920.1 8 - 9 1123 89
GenBank mRNA AJ006308.1 - 8 - 9 1123 89

View Full Product Details