Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB96
Entrez Gene ID
778073
Gene Name
MYB transcription factor MYB96
Gene Aliases
-
Chromosome Location
Chr.9: 37685598 - 37691370 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ822951.1 - 2 - 3 364 65

View Full Product Details