Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY14
Entrez Gene ID
100127371
Gene Name
WRKY14 protein
Gene Aliases
-
Chromosome Location
Chr.11: 3985019 - 3986672 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU019559.1 - 2 - 3 269 78

View Full Product Details