Gene Transcripts
- Gene Symbol
- MAPK14
- Entrez Gene ID
- 1432
- Gene Name
- mitogen-activated protein kinase 14
- Gene Aliases
- CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
- Chromosome Location
- Chr. 6: 36027635 - 36122964 on Build GRCh38
- UniGene ID
- Hs.485233
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001315.2 |
NP_001306.1
|
5, 6 |
921 |
|
NM_139012.2 |
NP_620581.1
|
5, 6 |
921 |
|
NM_139013.2 |
NP_620582.1
|
5, 6 |
921 |
|
NM_139014.2 |
NP_620583.1
|
5, 6 |
921 |
|
XM_006714998.3 |
XP_006715061.1
|
5, 6 |
396 |
|
XM_011514310.2 |
XP_011512612.2
|
5, 6 |
1189 |
|
XM_017010299.1 |
XP_016865788.1
|
5, 6 |
963 |
|
XM_017010300.1 |
XP_016865789.1
|
5, 6 |
963 |
|
XM_017010301.1 |
XP_016865790.1
|
5, 6 |
963 |
|
XM_017010302.1 |
XP_016865791.1
|
5, 6 |
963 |
|
XM_017010303.1 |
XP_016865792.1
|
5, 6 |
963 |
|
XM_017010304.1 |
XP_016865793.1
|
5, 6 |
963 |
|
GenBank mRNA |
AB074150.1 |
- |
5, 6 |
440 |
|
AF100544.1 |
- |
5, 6 |
455 |
|
AK291709.1 |
- |
5, 6 |
816 |
|
AK303414.1 |
- |
6, 7 |
485 |
|
BC000092.1 |
- |
5, 6 |
778 |
3505234 |
BC031574.1 |
- |
5, 6 |
777 |
5181064 |
BT006933.1 |
- |
5, 6 |
440 |
|
CR536505.1 |
- |
5, 6 |
440 |
|
FJ032367.1 |
- |
5, 6 |
440 |
|
FJ032368.1 |
- |
5, 6 |
440 |
|
L35263.1 |
- |
5, 6 |
802 |
|
L35264.1 |
- |
5, 6 |
648 |
|
View Full Product Details