Gene Transcripts

Gene Symbol
KRT3
Entrez Gene ID
3850
Gene Name
keratin 3
Gene Aliases
CK3, K3
Chromosome Location
Chr. 12: 52789685 - 52796108 on Build GRCh38
UniGene ID
Hs.680652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057088.2 NP_476429.2 4, 5 1089
GenBank mRNA AJ628418.1 - 4, 5 978
AK314987.1 - 4, 5 1098

View Full Product Details