Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195193.1 | NP_001182122.1 | 13 | 2305 | |
NM_002482.3 | NP_002473.2 | 15 | 2497 | ||
NM_152298.3 | NP_689511.2 | 14 | 1480 | ||
XM_005270888.2 | XP_005270945.1 | 16 | 2517 | ||
XM_005270889.2 | XP_005270946.1 | 15 | 1500 | ||
XM_011541509.1 | XP_011539811.1 | 15 | 2388 | ||
XM_017001356.1 | XP_016856845.1 | 13 | 1298 | ||
GenBank mRNA | AK092829.1 | - | 9 | 3518 | |
AK131249.1 | - | 10 | 2612 | ||
AK222778.1 | - | 15 | 2440 | ||
AK222816.1 | - | 15 | 2440 | ||
AK291637.1 | - | 15 | 2440 | ||
AK298893.1 | - | 13 | 2248 | ||
AK299584.1 | - | 14 | 2304 | ||
AL571577.2 | - | 1 | 752 | ||
BC003113.1 | - | 14 | 1398 | 3504220 | |
BC010105.2 | - | 15 | 2425 | 3538429 | |
BC011580.1 | - | 15 | 2423 | 4558913 | |
BC053608.1 | - | 14 | 1403 | 6013263 | |
BT006757.1 | - | 14 | 1321 |