Gene Transcripts
- Gene Symbol
- NDUFB5
- Entrez Gene ID
- 4711
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B5
- Gene Aliases
- CISGDH, SGDH
- Chromosome Location
- Chr. 3: 179604787 - 179624500 on Build GRCh38
- UniGene ID
- Hs.730674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199958.1 |
NP_001186887.1
|
3, 4 |
300 |
|
NM_002492.3 |
NP_002483.1
|
3, 4 |
300 |
|
GenBank mRNA |
AF047181.1 |
- |
3, 4 |
277 |
|
AK313822.1 |
- |
3, 4 |
278 |
|
AW955118.1 |
- |
3, 4 |
257 |
|
BC005271.1 |
- |
3, 4 |
291 |
3997377 |
BC009796.1 |
- |
3, 4 |
295 |
4284776 |
BC017909.1 |
- |
3, 4 |
290 |
|
BG776403.1 |
- |
3, 4 |
307 |
|
DA112751.1 |
- |
3, 4 |
303 |
|
View Full Product Details