Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000303.2 | NP_000294.1 | 4 | 354 | |
XM_005255374.4 | XP_005255431.1 | 3 | 277 | ||
GenBank mRNA | AK291537.1 | - | 4 | 325 | |
AK296123.1 | - | 4 | 347 | ||
AK299979.1 | - | 3 | 248 | ||
AK300785.1 | - | 4 | 330 | ||
AK311035.1 | - | 3 | 248 | ||
AK312532.1 | - | 4 | 325 | ||
BC008310.1 | - | 4 | 296 | 3611382 | |
DC366756.1 | - | 4 | 361 | ||
KU178265.1 | - | 4 | 284 | ||
U85773.1 | - | 4 | 332 |