Gene Transcripts
- Gene Symbol
- RASGRF1
- Entrez Gene ID
- 5923
- Gene Name
- Ras protein specific guanine nucleotide releasing factor 1
- Gene Aliases
- CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
- Chromosome Location
- Chr. 15: 78959947 - 79090873 on Build GRCh38
- UniGene ID
- Hs.459035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145648.1 |
NP_001139120.1
|
26, 27 |
4052 |
|
NM_002891.4 |
NP_002882.3
|
27, 28 |
4100 |
|
NM_153815.2 |
NP_722522.1
|
13, 14 |
2546 |
|
XM_011521867.2 |
XP_011520169.1
|
26, 27 |
3882 |
|
XM_017022455.1 |
XP_016877944.1
|
27, 28 |
3991 |
|
XM_017022456.1 |
XP_016877945.1
|
26, 27 |
3961 |
|
XM_017022459.1 |
XP_016877948.1
|
25, 26 |
3843 |
|
GenBank mRNA |
AK226101.1 |
- |
25, 26 |
3336 |
|
L26584.1 |
- |
27, 28 |
3731 |
|
M91815.1 |
- |
6, 7 |
593 |
|
S62035.1 |
- |
13, 14 |
2555 |
|
View Full Product Details