Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318798.1 | NP_001305727.1 | 18 | 2017 | |
NM_001318800.1 | NP_001305729.1 | 18 | 2301 | ||
NM_004186.4 | NP_004177.3 | 19 | 2278 | ||
XM_005265381.4 | XP_005265438.1 | 19 | 2408 | ||
XM_005265382.4 | XP_005265439.1 | 18 | 2118 | ||
XM_006713289.3 | XP_006713352.1 | 13 | 1890 | ||
XM_011533998.2 | XP_011532300.1 | 19 | 2210 | ||
GenBank mRNA | AB209259.1 | - | 18 | 1971 | |
BC042914.1 | - | 19 | 2274 | 5477898 | |
U33920.1 | - | 18 | 2154 | ||
U38276.1 | - | 18 | 2295 |