Gene Transcripts

Gene Symbol
MOGS
Entrez Gene ID
7841
Gene Name
mannosyl-oligosaccharide glucosidase
Gene Aliases
CDG2B, CWH41, DER7, GCS1
Chromosome Location
Chr. 2: 74461057 - 74465410 on Build GRCh38
UniGene ID
Hs.516119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146158.1 NP_001139630.1 2 148
NM_006302.2 NP_006293.2 1 474
XM_017004876.1 XP_016860365.1 1 464
XM_017004877.1 XP_016860366.1 2 139
GenBank mRNA AK292553.1 - 1 420
AK304655.1 - 1 426
AK307991.1 - 2 134
AK313355.1 - 1 474
BC028337.1 - 1 455 5110525
BC117250.1 - 1 347 40125801
BC117252.1 - 1 347 40125803
DC388420.1 - 2 120
X87237.1 - 1 443

View Full Product Details