Gene Transcripts

Gene Symbol
RRAGB
Entrez Gene ID
10325
Gene Name
Ras related GTP binding B
Gene Aliases
RAGB, bA465E19.1
Chromosome Location
Chr. X: 55717677 - 55760209 on Build GRCh38
UniGene ID
Hs.50282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006064.4 NP_006055.3 8 1455
NM_016656.3 NP_057740.2 9 1539
XM_005261973.4 XP_005262030.1 9 876
XM_017029217.1 XP_016884706.1 8 785
GenBank mRNA AK289407.1 - 8 1388
AL831926.1 - 8 1394
BC034726.1 - 8 1386 3919252
CD676755.1 - 2 180
X90530.1 - 9 1330

View Full Product Details