Gene Transcripts
- Gene Symbol
- TRDN
- Entrez Gene ID
- 10345
- Gene Name
- triadin
- Gene Aliases
- CPVT5, TDN, TRISK
- Chromosome Location
- Chr. 6: 123216339 - 123637093 on Build GRCh38
- UniGene ID
- Hs.144744
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001251987.1 |
NP_001238916.1
|
3, 4 |
701 |
|
NM_001256020.1 |
NP_001242949.1
|
3, 4 |
701 |
|
NM_001256021.1 |
NP_001242950.1
|
3, 4 |
701 |
|
NM_001256022.1 |
NP_001242951.1
|
3, 4 |
701 |
|
NM_006073.3 |
NP_006064.2
|
3, 4 |
701 |
|
GenBank mRNA |
AJ489257.2 |
- |
3, 4 |
596 |
|
AK291867.1 |
- |
3, 4 |
558 |
|
BC029393.1 |
- |
3, 4 |
558 |
|
BC057222.1 |
- |
3, 4 |
562 |
|
JN900469.1 |
- |
3, 4 |
515 |
|
U18985.1 |
- |
3, 4 |
405 |
|
View Full Product Details