Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
11137
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
IEF-SSP-9502
Chromosome Location
Chr. 12: 107685732 - 107713162 on Build GRCh38
UniGene ID
Hs.506652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317962.1 NP_001304891.1 6 596
NM_001317963.1 NP_001304892.1 6 768
NM_007062.2 NP_008993.1 6 740
GenBank mRNA AK223162.1 - 6 664
AK290681.1 - 6 581
AK296252.1 - 6 597
AK297961.1 - 5 425
BC000067.2 - 6 623
BC001652.2 - 6 659 2924215
BC010921.1 - 6 702
BC032127.2 - 6 662
BC040135.1 - 6 638
BC046144.1 - 6 654
L07758.1 - 6 659
Z36805.1 - 3 241

View Full Product Details