Gene Transcripts

Gene Symbol
C11orf24
Entrez Gene ID
53838
Gene Name
chromosome 11 open reading frame 24
Gene Aliases
DM4E3
Chromosome Location
Chr. 11: 68261335 - 68272001 on Build GRCh38
UniGene ID
Hs.303025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022338.3 NP_071733.1 4 1241
XM_005274053.3 XP_005274110.1 4 1416
XM_011545103.2 XP_011543405.1 2 836
GenBank mRNA AF264781.1 - 4 1201
AF370372.1 - 3 1109
AY358754.1 - 2 803
BC011765.2 - 3 1090 3614861

View Full Product Details