Gene Transcripts

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
Chromosome Location
Chr. 9: 19053137 - 19102942 on Build GRCh38
UniGene ID
Hs.533468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB046794.1 - 7 679
AF533709.1 - 14 2005
AF537091.1 - 14 1785
AL832495.1 - 14 1783
BC113981.1 - 14 1620 40082296

View Full Product Details