Gene Transcripts
- Gene Symbol
- TMEM63B
- Entrez Gene ID
- 55362
- Gene Name
- transmembrane protein 63B
- Gene Aliases
- C6orf110
- Chromosome Location
- Chr. 6: 44126628 - 44155519 on Build GRCh38
- UniGene ID
- Hs.414473
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318792.1 |
NP_001305721.1
|
6, 7 |
585 |
|
NM_018426.2 |
NP_060896.1
|
6, 7 |
551 |
|
XM_005249211.3 |
XP_005249268.1
|
8, 9 |
931 |
|
XM_005249213.4 |
XP_005249270.1
|
7, 8 |
858 |
|
XM_005249214.4 |
XP_005249271.1
|
5, 6 |
445 |
|
XM_005249215.4 |
XP_005249272.1
|
6, 7 |
750 |
|
XM_005249217.1 |
XP_005249274.1
|
6, 7 |
623 |
|
XM_006715135.1 |
XP_006715198.1
|
6, 7 |
607 |
|
XM_017010999.1 |
XP_016866488.1
|
8, 9 |
808 |
|
XM_017011000.1 |
XP_016866489.1
|
7, 8 |
789 |
|
XM_017011001.1 |
XP_016866490.1
|
5, 6 |
489 |
|
XM_017011002.1 |
XP_016866491.1
|
5, 6 |
592 |
|
GenBank mRNA |
AK093629.1 |
- |
6, 7 |
607 |
|
AK309791.1 |
- |
4, 5 |
454 |
|
BC022095.1 |
- |
3, 4 |
188 |
|
BC136769.1 |
- |
5, 6 |
427 |
9020759 |
CR627460.1 |
- |
6, 7 |
588 |
|
View Full Product Details