Gene Transcripts

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
Chromosome Location
Chr. 12: 4487730 - 4538508 on Build GRCh38
UniGene ID
Hs.302977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304811.1 NP_001291740.1 3 421
NM_020374.3 NP_065107.1 3 429
XM_005253716.1 XP_005253773.1 3 429
XM_006718992.3 XP_006719055.1 3 429
GenBank mRNA AJ272205.1 - 3 372
AK074479.1 - 3 429
AL834377.1 - 3 392
BC016908.1 - 3 372 4402263
BX641128.1 - 3 422
DA195770.1 - 3 394

View Full Product Details