Gene Transcripts

Gene Symbol
FN3K
Entrez Gene ID
64122
Gene Name
fructosamine 3 kinase
Gene Aliases
Chromosome Location
Chr. 17: 82735576 - 82751197 on Build GRCh38
UniGene ID
Hs.151135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022158.3 NP_071441.1 6 760
GenBank mRNA AJ404615.1 - 6 720
BC042680.1 - 6 704 5195472

View Full Product Details