Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_194279.3 | NP_919255.2 | 3 | 297 | |
GenBank mRNA | AK290728.1 | - | 3 | 270 | |
BC015771.1 | - | 3 | 257 | 4849790 | |
BC032893.2 | - | 3 | 252 | 5259721 | |
BX248252.1 | - | 3 | 255 |