Gene Transcripts

Gene Symbol
OR2T6
Entrez Gene ID
254879
Gene Name
olfactory receptor family 2 subfamily T member 6
Gene Aliases
OR2T6P, OR2T9, OST703
Chromosome Location
Chr. 1: 248387609 - 248388535 on Build GRCh38
UniGene ID
Hs.626884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005471.1 NP_001005471.1 1 491

View Full Product Details