Gene Transcripts

Gene Symbol
OR1L4
Entrez Gene ID
254973
Gene Name
olfactory receptor family 1 subfamily L member 4
Gene Aliases
OR1L5, OR9-29, OR9-E, OST046
Chromosome Location
Chr. 9: 122723990 - 122724925 on Build GRCh38
UniGene ID
Hs.626838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005235.1 NP_001005235.1 1 760

View Full Product Details