Gene Transcripts

Gene Symbol
OR5AN1
Entrez Gene ID
390195
Gene Name
olfactory receptor family 5 subfamily AN member 1
Gene Aliases
OR11-244
Chromosome Location
Chr. 11: 59364459 - 59365394 on Build GRCh38
UniGene ID
Hs.553755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004729.1 NP_001004729.1 1 559
GenBank mRNA BC140917.1 - 1 606

View Full Product Details