Gene Transcripts
- Gene Symbol
- TTYH1
- Entrez Gene ID
- 57348
- Gene Name
- tweety family member 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 54415431 - 54436719 on Build GRCh38
- UniGene ID
- Hs.268728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005367.2 |
NP_001005367.1
|
4, 5 |
758 |
|
NM_001201461.1 |
NP_001188390.1
|
4, 5 |
758 |
|
NM_020659.3 |
NP_065710.1
|
4, 5 |
758 |
|
XM_017027025.1 |
XP_016882514.1
|
4, 5 |
602 |
|
XM_017027026.1 |
XP_016882515.1
|
4, 5 |
602 |
|
GenBank mRNA |
AB162930.1 |
- |
4, 5 |
641 |
|
AB188496.1 |
- |
4, 5 |
758 |
|
AF177909.1 |
- |
4, 5 |
672 |
|
AK126690.1 |
- |
3, 4 |
669 |
|
AK289468.1 |
- |
4, 5 |
721 |
|
AK293468.1 |
- |
6, 7 |
845 |
|
BC011347.1 |
- |
4, 5 |
663 |
4215136 |
BX402889.2 |
- |
4, 5 |
738 |
|
View Full Product Details