Gene Transcripts

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
Chromosome Location
Chr. 2: 111766147 - 111884196 on Build GRCh38
UniGene ID
Hs.436527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022662.3 NP_073153.1 37 4742
XM_011511633.2 XP_011509935.1 37 4786
XM_011511634.2 XP_011509936.1 37 4901
XM_017004710.1 XP_016860199.1 37 4786
XM_017004711.1 XP_016860200.1 37 4774
XM_017004712.1 XP_016860201.1 37 4889
XM_017004713.1 XP_016860202.1 36 4849
XM_017004714.1 XP_016860203.1 36 4785
GenBank mRNA AJ278357.1 - 37 4757
AK023807.1 - 24 2968
AL832013.1 - 26 3098

View Full Product Details