Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080439.2 | NP_001073908.2 | 4 | 1437 | |
XM_011524283.1 | XP_011522585.1 | 4 | 1450 | ||
GenBank mRNA | BC033017.1 | - | 4 | 1401 | |
BC033020.1 | - | 4 | 1424 | 4823233 | |
BC115385.1 | - | 5 | 1013 | 40038134 | |
BC115386.1 | - | 5 | 1009 |