Gene Transcripts

Gene Symbol
SPATA31D5P
Entrez Gene ID
347127
Gene Name
SPATA31 subfamily D member 5, pseudogene
Gene Aliases
FAM75D5, FAM75D5P
Chromosome Location
Chr. 9: 81913437 - 81919927 on Build GRCh38
UniGene ID
Hs.215235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026851.1 4 523
GenBank mRNA AK125938.1 - 4 407
AK128229.1 - 3 1110
BC104780.1 - 4 443
BC104782.1 - 4 443

View Full Product Details