Gene Transcripts
- Gene Symbol
- PCYT1A
- Entrez Gene ID
- 5130
- Gene Name
- phosphate cytidylyltransferase 1, choline, alpha
- Gene Aliases
- CCTA, CT, CTA, CTPCT, PCYT1, SMDCRD
- Chromosome Location
- Chr. 3: 196237745 - 196287752 on Build GRCh38
- UniGene ID
- Hs.135997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001567.1 |
- |
1 |
474 |
|
- Gene Symbol
- ELAC2
- Entrez Gene ID
- 60528
- Gene Name
- elaC ribonuclease Z 2
- Gene Aliases
- COXPD17, ELC2, HPC2
- Chromosome Location
- Chr. 17: 12991612 - 13018064 on Build GRCh38
- UniGene ID
- Hs.434232
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094333.1 |
- |
1 |
367 |
|
- Gene Symbol
- LAMC3
- Entrez Gene ID
- 10319
- Gene Name
- laminin subunit gamma 3
- Gene Aliases
- OCCM
- Chromosome Location
- Chr. 9: 131009117 - 131093059 on Build GRCh38
- UniGene ID
- Hs.201805
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL133595.1 |
- |
6 |
3010 |
|
- Gene Symbol
- LINC00269
- Entrez Gene ID
- 100996279
- Gene Name
- long intergenic non-protein coding RNA 269
- Gene Aliases
- CXorf62, NCRNA00269
- Chromosome Location
- Chr. X: 69179557 - 69209924 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103715.1 |
|
5 |
688 |
|
GenBank mRNA |
AK090929.1 |
- |
5 |
688 |
|
- Gene Symbol
- SERPINB6
- Entrez Gene ID
- 5269
- Gene Name
- serpin family B member 6
- Gene Aliases
- CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3
- Chromosome Location
- Chr. 6: 2948159 - 2972165 on Build GRCh38
- UniGene ID
- Hs.519523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK122895.1 |
- |
1 |
110 |
|
AK122961.1 |
- |
1 |
958 |
|
AK128637.1 |
- |
1 |
1538 |
|
View Full Product Details