Gene Transcripts
- Gene Symbol
- CYP4F30P
- Entrez Gene ID
- 100132708
- Gene Name
- cytochrome P450 family 4 subfamily F member 30, pseudogene
- Gene Aliases
- C2orf14
- Chromosome Location
- Chr. 2: 130680050 - 130685863 on Build GRCh38
- UniGene ID
- Hs.734862
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093281.1 |
- |
5 |
1723 |
|
- Gene Symbol
- FAR2P1
- Entrez Gene ID
- 440905
- Gene Name
- fatty acyl-CoA reductase 2 pseudogene 1
- Gene Aliases
- HEL-182
- Chromosome Location
- Chr. 2: 130025998 - 130051131 on Build GRCh38
- UniGene ID
- Hs.469918
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL043002.1 |
- |
4 |
591 |
|
View Full Product Details