Gene Transcripts

Gene Symbol
ZGLP1
Entrez Gene ID
100125288
Gene Name
zinc finger, GATA-like protein 1
Gene Aliases
GATAD3, GLP-1, GLP1
Chromosome Location
Chr. 19: 10304803 - 10309557 on Build GRCh38
UniGene ID
Hs.709296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103167.1 NP_001096637.1 1 1162
GenBank mRNA AK096830.1 - 1 1485

View Full Product Details