Gene Transcripts

Gene Symbol
PDAP1
Entrez Gene ID
11333
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
Chromosome Location
Chr. 7: 99394673 - 99408682 on Build GRCh38
UniGene ID
Hs.632296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014891.6 NP_055706.1 5 530
GenBank mRNA BC000684.2 - 5 442
BC007873.2 - 5 406 4303959
BC071993.1 - 5 440
BG281467.1 - 5 528
BI460344.1 - 5 522
U41745.1 - 5 417

View Full Product Details