Gene Transcripts

Gene Symbol
HMX3
Entrez Gene ID
340784
Gene Name
H6 family homeobox 3
Gene Aliases
NKX-5.1, NKX5.1, Nkx5-1
Chromosome Location
Chr. 10: 123136051 - 123137731 on Build GRCh38
UniGene ID
Hs.531194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105574.1 NP_001099044.1 2 516
GenBank mRNA AW088771.1 - 1 31

View Full Product Details