Gene Transcripts

Gene Symbol
IER5L
Entrez Gene ID
389792
Gene Name
immediate early response 5 like
Gene Aliases
bA247A12.2
Chromosome Location
Chr. 9: 129175552 - 129178261 on Build GRCh38
UniGene ID
Hs.529857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203434.2 NP_982258.2 1 1409
GenBank mRNA BC064028.1 - 2 1040
BQ574938.1 - 1 206

View Full Product Details