Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002639.4 | NP_002630.2 | 2, 3 | 304 | |
GenBank mRNA | AK312765.1 | - | 2, 3 | 282 | |
AL602439.1 | - | 2, 3 | 307 | ||
BC020713.1 | - | 2, 3 | 257 | 4248249 | |
U04313.1 | - | 2, 3 | 237 |