Gene Transcripts
- Gene Symbol
- EPB41L4B
- Entrez Gene ID
- 54566
- Gene Name
- erythrocyte membrane protein band 4.1 like 4B
- Gene Aliases
- CG1, EHM2
- Chromosome Location
- Chr. 9: 109171974 - 109320964 on Build GRCh38
- UniGene ID
- Hs.591901
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018424.3 |
NP_060894.2
|
4, 5 |
1048 |
|
NM_019114.4 |
NP_061987.3
|
4, 5 |
1048 |
|
XM_011518790.1 |
XP_011517092.1
|
4, 5 |
545 |
|
XM_011518791.2 |
XP_011517093.1
|
4, 5 |
530 |
|
XM_011518792.2 |
XP_011517094.1
|
4, 5 |
530 |
|
XM_011518794.2 |
XP_011517096.1
|
4, 5 |
530 |
|
XM_017014813.1 |
XP_016870302.1
|
4, 5 |
530 |
|
XM_017014814.1 |
XP_016870303.1
|
4, 5 |
530 |
|
XM_017014815.1 |
XP_016870304.1
|
4, 5 |
530 |
|
GenBank mRNA |
AB032179.3 |
- |
4, 5 |
1048 |
|
AF153416.1 |
- |
4, 5 |
820 |
|
AF153418.1 |
- |
4, 5 |
820 |
|
AK000388.1 |
- |
4, 5 |
432 |
|
View Full Product Details