The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031847.2 | NP_001027017.1 | 3 - 4 | 457 | 85 |
NM_001876.3 | NP_001867.2 | 3 - 4 | 457 | 85 | |
XM_005273762.2 | XP_005273819.1 | 3 - 4 | 433 | 85 | |
XM_005273763.1 | XP_005273820.1 | 3 - 4 | 433 | 85 | |
XM_017017220.1 | XP_016872709.1 | 3 - 4 | 566 | 85 | |
GenBank mRNA | AK172798.1 | - | 3 - 4 | 443 | 85 |
AK309464.1 | - | 3 - 4 | 433 | 85 | |
AK314301.1 | - | 3 - 4 | 396 | 85 | |
BC000185.1 | - | 3 - 4 | 332 | 85 | |
BT009791.1 | - | 2 - 3 | 287 | 85 | |
DB063502.1 | - | 3 - 4 | 457 | 85 | |
L39211.1 | - | 3 - 4 | 363 | 85 |