Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLN
Entrez Gene ID
5350
Gene Name
phospholamban
Gene Aliases
CMD1P, CMH18, PLB
Chromosome Location
Chr.6: 118548263 - 118561715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002667.4 NP_002658.1 1 - 2 130 98
GenBank mRNA AK311799.1 - 1 - 2 65 98
BC005269.1 - 1 - 2 65 98
BP257094.1 - 1 - 2 130 98
BP259965.1 - 1 - 2 114 98
M63603.1 - 1 - 2 84 98

View Full Product Details