The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002788.3 | NP_002779.1 | 5 - 6 | 517 | 99 |
NR_038123.1 | 4 - 5 | 434 | 99 | ||
GenBank mRNA | AK315158.1 | - | 5 - 6 | 466 | 99 |
BC029402.1 | - | 5 - 6 | 447 | 99 | |
BC038990.1 | - | 5 - 6 | 444 | 99 | |
BG703488.1 | - | 4 - 5 | 340 | 99 | |
BT019715.1 | - | 5 - 6 | 401 | 99 | |
CF128376.1 | - | 5 - 6 | 517 | 99 | |
CR456933.1 | - | 5 - 6 | 401 | 99 | |
D00762.1 | - | 5 - 6 | 406 | 99 |