The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303029.1 | NP_001289958.1 | 2 - 3 | 307 | 117 |
NM_007183.3 | NP_009114.1 | 1 - 2 | 343 | 117 | |
GenBank mRNA | AF053719.1 | - | 1 - 2 | 287 | 117 |
AK025067.1 | - | 1 - 2 | 304 | 117 | |
AK223511.1 | - | 1 - 2 | 304 | 117 | |
BC000081.2 | - | 1 - 2 | 309 | 117 | |
BX356582.2 | - | 2 - 3 | 262 | 117 | |
DB139782.1 | - | 1 - 2 | 343 | 117 | |
FN421477.1 | - | 2 - 3 | 307 | 117 | |
Z98265.1 | - | 1 - 2 | 307 | 117 |